siRNA Details
Gene Symbol: Acvr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031571.2 | NP_113759.1 | 7 | 915 | ||
XM_008761703.1 | XP_008759925.1 | 8 | 994 | |||
GenBank mRNA | L10639.1 | - | 7 | 898 | ||
S48190.1 | - | 7 | 915 |
