siRNA Details
| Gene Symbol: Slc1a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175758.3 | NP_786934.1 | 8 | 1801 | ||
| GenBank mRNA | AJ132846.1 | - | 8 | 1532 | ||
| BC080242.1 | - | 8 | 1801 | 7100054 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc1a5| Gene Symbol: Slc1a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175758.3 | NP_786934.1 | 8 | 1801 | ||
| GenBank mRNA | AJ132846.1 | - | 8 | 1532 | ||
| BC080242.1 | - | 8 | 1801 | 7100054 | ||