siRNA Details
| Gene Symbol: Ccdc61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106229.1 | NP_001099699.1 | 4, 5 | 724 | ||
| GenBank mRNA | BC127532.1 | - | 4, 5 | 626 | 7132439 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc61| Gene Symbol: Ccdc61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106229.1 | NP_001099699.1 | 4, 5 | 724 | ||
| GenBank mRNA | BC127532.1 | - | 4, 5 | 626 | 7132439 | |