siRNA Details
| Gene Symbol: Sympk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100830.1 | NP_001094300.1 | 7 | 610 | ||
| XM_008758908.1 | XP_008757130.1 | 7 | 860 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sympk| Gene Symbol: Sympk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100830.1 | NP_001094300.1 | 7 | 610 | ||
| XM_008758908.1 | XP_008757130.1 | 7 | 860 | |||