siRNA Details
| Gene Symbol: Rsph6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024748.1 | NP_001019919.1 | 4 | 1709 | ||
| XM_008758909.1 | XP_008757131.1 | 4 | 1764 | |||
| GenBank mRNA | BC097421.1 | - | 4 | 1709 | 7454827 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rsph6a| Gene Symbol: Rsph6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024748.1 | NP_001019919.1 | 4 | 1709 | ||
| XM_008758909.1 | XP_008757131.1 | 4 | 1764 | |||
| GenBank mRNA | BC097421.1 | - | 4 | 1709 | 7454827 | |