siRNA Details
| Gene Symbol: Qpctl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106230.1 | NP_001099700.1 | 5 | 893 | ||
| XM_006228402.2 | XP_006228464.1 | 5 | 1335 | |||
| GenBank mRNA | BC169076.1 | - | 5 | 889 | 5598782 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Qpctl| Gene Symbol: Qpctl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106230.1 | NP_001099700.1 | 5 | 893 | ||
| XM_006228402.2 | XP_006228464.1 | 5 | 1335 | |||
| GenBank mRNA | BC169076.1 | - | 5 | 889 | 5598782 | |