siRNA Details
| Gene Symbol: RGD1307554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109664.1 | NP_001103134.1 | 5 | 358 | ||
| XM_006228571.1 | XP_006228633.1 | 5 | 374 | |||
| XM_006228572.2 | XP_006228634.2 | 5 | 374 | |||
| XM_008759118.1 | XP_008757340.1 | 4 | 644 | |||
| GenBank mRNA | BC127543.1 | - | 5 | 343 | 7314257 | |

RGD1307554
Genomic Map