siRNA Details
| Gene Symbol: Plekhg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134972.1 | NP_001128444.1 | 19 | 3903 | ||
| XM_006228613.2 | XP_006228675.1 | 17 | 3749 | |||
| XM_006228614.1 | XP_006228676.1 | 18 | 3656 | |||
| XM_006228615.2 | XP_006228677.1 | 19 | 4592 | |||
| XM_006228616.2 | XP_006228678.1 | 19 | 3345 | |||
| XM_006228617.2 | XP_006228679.1 | 18 | 3392 | |||
| XM_006228618.2 | XP_006228680.1 | 18 | 3389 | |||
| XM_006228619.2 | XP_006228681.1 | 18 | 3347 | |||
| XM_006228620.2 | XP_006228682.1 | 18 | 3344 | |||

Plekhg2
Genomic Map