siRNA Details
Gene Symbol: Sars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106240.1 | NP_001099710.1 | 1 | 146 | ||
GenBank mRNA | FQ212522.1 | - | 1 | 148 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106240.1 | NP_001099710.1 | 1 | 146 | ||
GenBank mRNA | FQ212522.1 | - | 1 | 148 |