siRNA Details
| Gene Symbol: Rinl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106241.1 | NP_001099711.1 | 10, 11 | 1606 | ||
| XM_006228653.2 | XP_006228715.1 | 10, 11 | 1858 | |||
| XM_006228654.2 | XP_006228716.1 | 10, 11 | 1846 | |||
| XM_006228657.2 | XP_006228719.1 | 9, 10 | 1581 | |||
| XM_008759120.1 | XP_008757342.1 | 9, 10 | 1723 | |||
| XM_008759121.1 | XP_008757343.1 | 10, 11 | 1691 | |||

Rinl
Genomic Map