siRNA Details
| Gene Symbol: Rinl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106241.1 | NP_001099711.1 | 2, 3 | 154 | ||
| XM_006228653.2 | XP_006228715.1 | 2, 3 | 406 | |||
| XM_006228654.2 | XP_006228716.1 | 2, 3 | 394 | |||
| XM_006228655.2 | XP_006228717.1 | 2, 3 | 406 | |||
| XM_006228656.2 | XP_006228718.1 | 2, 3 | 406 | |||
| XM_008759121.1 | XP_008757343.1 | 2, 3 | 239 | |||

Rinl
Genomic Map