siRNA Details
| Gene Symbol: Catsperg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170340.1 | NP_001163811.1 | 17 | 1943 | ||
| XM_006228722.2 | XP_006228784.1 | 17 | 2529 | |||
| XM_008759124.1 | XP_008757346.1 | 17 | 2562 | |||
| XM_008759125.1 | XP_008757347.1 | 17 | 2544 | |||
| XM_008759126.1 | XP_008757348.1 | 16 | 2454 | |||
| XM_008759127.1 | XP_008757349.1 | 16 | 1760 | |||
| GenBank mRNA | BC167101.1 | - | 10 | 956 | ||

Catsperg1
Genomic Map