siRNA Details
| Gene Symbol: Catsperg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170340.1 | NP_001163811.1 | 14 | 1590 | ||
| XM_006228722.2 | XP_006228784.1 | 14 | 2176 | |||
| XM_008759124.1 | XP_008757346.1 | 14 | 2209 | |||
| XM_008759125.1 | XP_008757347.1 | 14 | 2191 | |||
| XM_008759126.1 | XP_008757348.1 | 13 | 2101 | |||
| XM_008759127.1 | XP_008757349.1 | 13 | 1407 | |||
| GenBank mRNA | BC167101.1 | - | 7 | 603 | ||

Catsperg1
Genomic Map