siRNA Details
| Gene Symbol: Tbcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040180.1 | NP_001035270.1 | 3 | 561 | ||
| GenBank mRNA | AB256046.1 | - | 3 | 561 | ||
| AM258961.1 | - | 3 | 561 | |||
| BC126061.1 | - | 3 | 379 | 7113234 | ||
| DQ447885.1 | - | 3 | 360 | |||
| Gene Symbol: LOC103690005 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759264.1 | XP_008757486.1 | 3 | 391 | ||

Tbcb
Genomic Map