siRNA Details
| Gene Symbol: Lin37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106245.1 | NP_001099715.1 | 6, 7 | 627 | ||
| XM_006228769.2 | XP_006228831.1 | 6, 7 | 1048 | |||
| GenBank mRNA | FQ233946.1 | - | 6, 7 | 595 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lin37| Gene Symbol: Lin37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106245.1 | NP_001099715.1 | 6, 7 | 627 | ||
| XM_006228769.2 | XP_006228831.1 | 6, 7 | 1048 | |||
| GenBank mRNA | FQ233946.1 | - | 6, 7 | 595 | ||