siRNA Details
| Gene Symbol: Pop4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009642.1 | NP_001009642.1 | 5 | 552 | ||
| GenBank mRNA | BC088183.1 | - | 5 | 552 | 7365498 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pop4| Gene Symbol: Pop4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009642.1 | NP_001009642.1 | 5 | 552 | ||
| GenBank mRNA | BC088183.1 | - | 5 | 552 | 7365498 | |