siRNA Details
| Gene Symbol: Siglec5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106249.2 | NP_001099719.2 | 2 | 307 | ||
| XM_006228946.2 | XP_006229008.1 | 2 | 420 | |||
| XM_008759327.1 | XP_008757549.1 | 2 | 395 | |||
| XM_008759328.1 | XP_008757550.1 | 2 | 395 | |||
| XM_008759329.1 | XP_008757551.1 | 2 | 397 | |||

Siglec5
Genomic Map