siRNA Details
| Gene Symbol: LOC102553035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229178.2 | XP_006229240.2 | 2 | 207 | ||
| XM_008759447.1 | XP_008757669.1 | 3 | 187 | |||
| Gene Symbol: Klk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728759.4 | XP_002728805.2 | 3 | 352 | ||

LOC102553035
Genomic Map