siRNA Details
| Gene Symbol: Zfp473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229185.1 | XP_006229247.1 | 5 | 2779 | ||
| XM_006229186.1 | XP_006229248.1 | 5 | 2397 | |||
| XM_006229187.1 | XP_006229249.1 | 5 | 2350 | |||
| XM_006229188.2 | XP_006229250.1 | 5 | 2278 | |||
| XM_008759453.1 | XP_008757675.1 | 4 | 2602 | |||
| XM_008759454.1 | XP_008757676.1 | 4 | 2146 | |||

Zfp473
Genomic Map