siRNA Details
| Gene Symbol: Zfp473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229185.1 | XP_006229247.1 | 2 | 647 | ||
| XM_006229186.1 | XP_006229248.1 | 2 | 265 | |||
| XM_006229187.1 | XP_006229249.1 | 2 | 218 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp473| Gene Symbol: Zfp473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229185.1 | XP_006229247.1 | 2 | 647 | ||
| XM_006229186.1 | XP_006229248.1 | 2 | 265 | |||
| XM_006229187.1 | XP_006229249.1 | 2 | 218 | |||