siRNA Details
| Gene Symbol: Irf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006969.1 | NP_001006970.1 | 6 | 1066 | ||
| XM_006229021.1 | XP_006229083.1 | 6 | 1668 | |||
| XM_006229022.2 | XP_006229084.1 | 6 | 1628 | |||
| XM_006229023.2 | XP_006229085.1 | 6 | 1187 | |||
| XM_006229024.2 | XP_006229086.1 | 6 | 1055 | |||
| XM_008759337.1 | XP_008757559.1 | 5 | 1016 | |||
| GenBank mRNA | BC083776.1 | - | 6 | 1066 | 7193483 | |

Irf3
Genomic Map