siRNA Details
| Gene Symbol: Fam83e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_218559.8 | XP_218559.4 | 2 | 1680 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam83e| Gene Symbol: Fam83e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_218559.8 | XP_218559.4 | 2 | 1680 | ||