siRNA Details
| Gene Symbol: Reps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106264.1 | NP_001099734.1 | 1478 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Reps1| Gene Symbol: Reps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106264.1 | NP_001099734.1 | 1478 | |||