siRNA Details
Gene Symbol: LOC100909392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758664.1 | XP_008756886.1 | 7 | 1264 |
Gene Symbol: Reps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106264.1 | NP_001099734.1 | 2205 |
