siRNA Details
| Gene Symbol: Nxf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037216.1 | NP_001032293.1 | 20 | 1874 | ||
| XM_006257254.2 | XP_006257316.1 | 20 | 1907 | |||
| XM_008773431.1 | XP_008771653.1 | 20 | 2445 | |||
| GenBank mRNA | BC100151.1 | - | 20 | 1874 | 7456178 | |
| FQ229456.1 | - | 9 | 701 | |||

Nxf7
Genomic Map