siRNA Details
| Gene Symbol: Zfp592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106272.1 | NP_001099742.1 | 2 | 610 | ||
| XM_006229424.2 | XP_006229486.1 | 2 | 768 | |||
| XM_006229425.2 | XP_006229487.1 | 2 | 768 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp592| Gene Symbol: Zfp592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106272.1 | NP_001099742.1 | 2 | 610 | ||
| XM_006229424.2 | XP_006229486.1 | 2 | 768 | |||
| XM_006229425.2 | XP_006229487.1 | 2 | 768 | |||