siRNA Details
| Gene Symbol: Rhcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183053.1 | NP_898876.1 | 3 | 504 | ||
| GenBank mRNA | AY129073.1 | - | 3 | 504 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhcg| Gene Symbol: Rhcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183053.1 | NP_898876.1 | 3 | 504 | ||
| GenBank mRNA | AY129073.1 | - | 3 | 504 | ||