siRNA Details
| Gene Symbol: Abhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106275.1 | NP_001099745.1 | 3 | 384 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abhd2| Gene Symbol: Abhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106275.1 | NP_001099745.1 | 3 | 384 | ||