siRNA Details
| Gene Symbol: Olr20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000118.1 | NP_001000118.1 | 1 | 496 | ||
| Gene Symbol: LOC100911035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752172.2 | XP_003752220.2 | 1 | 496 | ||

Olr20
Genomic Map