siRNA Details
| Gene Symbol: Tmem126b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106280.2 | NP_001099750.2 | 2, 3 | 201 | ||
| GenBank mRNA | BC167111.1 | - | 2, 3 | 201 | 6921139 | |
| FQ223509.1 | - | 2, 3 | 202 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem126b| Gene Symbol: Tmem126b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106280.2 | NP_001099750.2 | 2, 3 | 201 | ||
| GenBank mRNA | BC167111.1 | - | 2, 3 | 201 | 6921139 | |
| FQ223509.1 | - | 2, 3 | 202 | |||