siRNA Details
| Gene Symbol: Ankrd42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135013.1 | NP_001128485.1 | 3, 4 | 1255 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd42| Gene Symbol: Ankrd42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135013.1 | NP_001128485.1 | 3, 4 | 1255 | ||