siRNA Details
Gene Symbol: Micalcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182669.2 | NP_872610.2 | 8 | 1911 | ||
GenBank mRNA | AY149342.1 | - | 5 | 1351 | ||
BC098647.1 | - | 8 | 1911 | 7112444 | ||
BC100137.1 | - | 5 | 1365 | 7457561 |
