siRNA Details
Gene Symbol: Gif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017162.3 | NP_058858.1 | 5 | 665 | ||
GenBank mRNA | J03577.1 | - | 5 | 666 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017162.3 | NP_058858.1 | 5 | 665 | ||
GenBank mRNA | J03577.1 | - | 5 | 666 |