siRNA Details
| Gene Symbol: Olr106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000147.1 | NP_001000147.1 | 1 | 397 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr106| Gene Symbol: Olr106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000147.1 | NP_001000147.1 | 1 | 397 | ||