siRNA Details
| Gene Symbol: Capza3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017164.2 | NP_058860.2 | 1 | 371 | ||
| GenBank mRNA | BC078870.1 | - | 1 | 371 | 7133258 | |
| Y12538.1 | - | 1 | 351 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Capza3| Gene Symbol: Capza3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017164.2 | NP_058860.2 | 1 | 371 | ||
| GenBank mRNA | BC078870.1 | - | 1 | 371 | 7133258 | |
| Y12538.1 | - | 1 | 351 | |||