siRNA Details
Gene Symbol: Capza3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017164.2 | NP_058860.2 | 1 | 723 | ||
GenBank mRNA | BC078870.1 | - | 1 | 723 | 7133258 | |
Y12538.1 | - | 1 | 703 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Capza3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017164.2 | NP_058860.2 | 1 | 723 | ||
GenBank mRNA | BC078870.1 | - | 1 | 723 | 7133258 | |
Y12538.1 | - | 1 | 703 |