siRNA Details
Gene Symbol: Olr122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000156.1 | NP_001000156.1 | 1 | 287 |
Gene Symbol: LOC100361642 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229925.1 | XP_006229987.1 | 1 | 287 |
