siRNA Details
| Gene Symbol: Olr155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000168.1 | NP_001000168.1 | 1 | 483 | ||
| XM_006229911.2 | XP_006229973.2 | 2 | 1684 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr155| Gene Symbol: Olr155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000168.1 | NP_001000168.1 | 1 | 483 | ||
| XM_006229911.2 | XP_006229973.2 | 2 | 1684 | |||