siRNA Details
Gene Symbol: Olr227 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000203.1 | NP_001000203.1 | 1 | 781 |
Gene Symbol: LOC100912097 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748913.1 | XP_003748961.1 | 1 | 781 |
