siRNA Details
| Gene Symbol: Olr233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000207.1 | NP_001000207.1 | 1 | 927 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr233| Gene Symbol: Olr233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000207.1 | NP_001000207.1 | 1 | 927 | ||