siRNA Details
Gene Symbol: Hbs1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011934.1 | NP_001011934.1 | 15 | 1821 | ||
GenBank mRNA | BC079463.1 | - | 15 | 1821 | 7122195 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hbs1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011934.1 | NP_001011934.1 | 15 | 1821 | ||
GenBank mRNA | BC079463.1 | - | 15 | 1821 | 7122195 |