siRNA Details
| Gene Symbol: Prkce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017171.1 | NP_058867.1 | 9 | 1594 | ||
| GenBank mRNA | AY642593.1 | - | 9 | 1408 | ||
| M15523.1 | - | 2 | 214 | |||
| M18331.1 | - | 9 | 1594 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkce| Gene Symbol: Prkce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017171.1 | NP_058867.1 | 9 | 1594 | ||
| GenBank mRNA | AY642593.1 | - | 9 | 1408 | ||
| M15523.1 | - | 2 | 214 | |||
| M18331.1 | - | 9 | 1594 | |||