siRNA Details
| Gene Symbol: Prkce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017171.1 | NP_058867.1 | 14 | 2312 | ||
| GenBank mRNA | AY642593.1 | - | 14 | 2126 | ||
| M18331.1 | - | 14 | 2312 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkce| Gene Symbol: Prkce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017171.1 | NP_058867.1 | 14 | 2312 | ||
| GenBank mRNA | AY642593.1 | - | 14 | 2126 | ||
| M18331.1 | - | 14 | 2312 | |||