siRNA Details
| Gene Symbol: Olr282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000224.1 | NP_001000224.1 | 1 | 298 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr282| Gene Symbol: Olr282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000224.1 | NP_001000224.1 | 1 | 298 | ||