siRNA Details
| Gene Symbol: Eif3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277302.1 | NP_001264231.1 | 2, 3 | 476 | ||
| GenBank mRNA | FQ212642.1 | - | 2, 3 | 465 | ||
| FQ213714.1 | - | 2, 3 | 476 | |||
| FQ229379.1 | - | 2, 3 | 150 | |||
| FQ230130.1 | - | 2, 3 | 464 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_025344, BC070473, AK002778, AK190701, AK197956, and others.

Eif3f
Genomic Map