siRNA Details
| Gene Symbol: Eif3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277302.1 | NP_001264231.1 | 7 | 1004 | ||
| GenBank mRNA | FQ212642.1 | - | 7 | 993 | ||
| FQ213714.1 | - | 7 | 1005 | |||
| FQ219431.1 | - | 2 | 197 | |||
| FQ229379.1 | - | 7 | 678 | |||
| FQ230130.1 | - | 7 | 992 | |||

Eif3f
Genomic Map