siRNA Details
| Gene Symbol: Eif3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277302.1 | NP_001264231.1 | 4 | 645 | ||
| GenBank mRNA | FQ212642.1 | - | 4 | 634 | ||
| FQ230130.1 | - | 4 | 633 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif3f| Gene Symbol: Eif3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277302.1 | NP_001264231.1 | 4 | 645 | ||
| GenBank mRNA | FQ212642.1 | - | 4 | 634 | ||
| FQ230130.1 | - | 4 | 633 | |||