siRNA Details
Gene Symbol: Uqcrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006970.1 | NP_001006971.1 | 12 | 1096 | ||
GenBank mRNA | BC083610.1 | - | 12 | 1096 | 7133503 | |
FQ219943.1 | - | 4 | 377 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Uqcrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006970.1 | NP_001006971.1 | 12 | 1096 | ||
GenBank mRNA | BC083610.1 | - | 12 | 1096 | 7133503 | |
FQ219943.1 | - | 4 | 377 |