siRNA Details
| Gene Symbol: Ubfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034911.1 | NP_001030083.1 | 3 | 467 | ||
| XM_006230189.2 | XP_006230251.1 | 3 | 740 | |||
| GenBank mRNA | BC101866.1 | - | 3 | 467 | 7938969 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubfd1| Gene Symbol: Ubfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034911.1 | NP_001030083.1 | 3 | 467 | ||
| XM_006230189.2 | XP_006230251.1 | 3 | 740 | |||
| GenBank mRNA | BC101866.1 | - | 3 | 467 | 7938969 | |