siRNA Details
| Gene Symbol: LOC103694867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773816.1 | XP_008772038.1 | 12 | 1805 | ||
| Gene Symbol: Gga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100519.1 | NP_001093989.1 | 13 | 1299 | ||
| GenBank mRNA | BC098781.1 | - | 7 | 577 | ||

LOC103694867
Genomic Map